Learn More
Essendant GLOVESNITRL12XL10WH
Supplier:  Essendant ANS56886
Encompass_Preferred
Kimtech is your partner in driving progress As part of Kimberly Clark s 150-year-old legacy of innovation and partnership with the scientific community we re proud to offer Kimtech G3 Nitrile Gloves When you need a reliable cleanroom glove Kimtech G3 Nitrile Gloves are a smart choice Kimtech G3 Nitrile Gloves provide enhanced cleanroom protection against a wide range of contaminants chemicals viruses and cytotoxic drugs These non-sterile cleanroom gloves offer a comfortable fit and feature textured fingertips for a secure grip excellent dexterity and tactile sensitivity These ambidextrous gloves have textured fingertips are double donnable and have a smooth finish They feature a beaded cuff to help prevent roll-down These gloves have an AQL of 1 5 are anti-static tested and are not made with natural rubber latex (latex and silicon free) Additionally these gloves perform with a cleanliness level of a maximum of 950 particles gt/0 5 m/cm2 These nitrile gloves are envir
Catalog No. 50-000-23068