Learn More
Essendant GLOVESNTRLXTRAL1KWH
Supplier:  Essendant ANS53140
Encompass_Preferred
Kimtech is your partner in driving progress As part of Kimberly Clark s 150-year-old legacy of innovation and partnership with the scientific community we re proud to offer Kimtech Sterling Nitrile Exam Gloves These are low-cost nitrile gloves combining the sensitivity of latex with the protection and strength of nitrile The innovative gloves manufacturing technology delivers a comfortable and affordable glove with the protection and cleanliness you need The gloves are available are ambidextrous powder-free have an AQL of 1 0 and feature beaded cuffs and textured fingertips Glove Material- Nitrile Packaged Sterile- No Size Type- Large Length- 12
Catalog No. 50-000-20351