Learn More
Essendant GLOVESNYLON9LRD
Supplier:  Essendant NSPNF119L
Encompass_Preferred
Gloves are made with a soft and flexible foamed PVC coating which provides shock and vibration absorption for the wearer Stretchy and form-fitting seamless nylon liners create a snug fit for exceptional comfort and reduced hand fatigue PVC coating is tough as well as comfortable with high abrasion resistance for extra protection Foamed coating provides an open-surface texture which clears liquids from the surface and gives the wearer a strong wet grip The uncoated back creates ventilation and increases wearer comfort Gloves are launderable and have a long service life Anti-Vibration- No EN 388 Badge- 4131 Size Type- Large Size Number- Size 9
Catalog No. 50-000-33604