Learn More
Essendant GLOVESPU 9277PUMLIM
Supplier:  Essendant CRW9277PUM
Encompass_Preferred
The MCR Safety Cut Pro series offers various cut protection levels with multiple color and coating options The 9277PU cut-resistant gloves feature a 13-gauge fluorescent lime HyperMax shell They are coated on the palm and fingertips with a black PU (polyurethane) coating which provides better abrasion resistance and enhanced grip The fluorescent lime color adds another layer of protection making the wearer more visible ANSI Cut Resistance- A7 ANSI Abrasion Resistance- Level 6 ANSI Puncture Resistance- Level 4 Anti-Vibration- No
Catalog No. 50-000-20273