Learn More
Essendant GLOVESSEAMLSS DIPSMLBE
Supplier:  Essendant CRW96731SDZ
Encompass_Preferred
Multipurpose work gloves provide durable strength and grip without compromising breathability Gray nylon shell offers added comfort and flexibility Blue foam latex-dipped palm and fingertips maintain a solid grip in both wet and dry environments Ideal for small parts handling where tactile sensitivity and dexterity are important Memphis Flex Latex-Coated Seamless Nylon Knit ANSI Abrasion Resistance- Level 3 ANSI Puncture Resistance- Level 2 Anti-Vibration- No EN 388 Badge- 3131
Catalog No. 50-000-21354