Learn More
Essendant GLOVESSEEMLSS DIPXLGGY
Supplier:  Essendant CRW9674XLDZ
Encompass_Preferred
Strength and comfort are a durable combination with these nitrile-dipped work gloves Ultra Tech foam provides additional protection against heavier tasks and cushions impacts Nitrile-dipped palm and fingers maintain a solid grip during both wet and dry applications Seamless nylon knit design offers flexibility Ideal for small parts handling where tactile sensitivity and dexterity are important ANSI Abrasion Resistance- Level 4 ANSI Puncture Resistance- Level 2 Anti-Vibration- No EN 388 Badge- 3121
Catalog No. 50-000-19433