Learn More
Essendant GLOVESSUPR DUTYXLBK/YL
Supplier:  Essendant IRNSDG205XL
Encompass_Preferred
Trust Advanced Glove Technology (AGT) to keep your hands safe Extensive Exo-Guard knuckle armor and synthetic leather sidewalls provide flexible finger-impact protection Duraclad reinforcement points enhance abrasion resistance TPR metacarpal protection shields the back of your hand Hybrid cuff system with TPR puller delivers a snug comfortable fit while making it easier to put the gloves on and take them off Handy terry-cloth perspiration wipe is conveniently integrated into the back of the thumb Anti-Vibration- No EN 388 Badge- 4132 Size Type- X-Large Color(s)- Black/Yellow
Catalog No. 50-000-32273