Learn More
Essendant GLOVESSYNTHETICPFMCRE
Supplier:  Essendant BWK315MBX
Encompass_Preferred
Sturdy synthetic vinyl gloves give the look and feel of latex without the latex allergens They offer exceptional strength and protection yet maintain maximum tactile sensitivity This glove is designed as the ultimate solution for those who are at risk of becoming sensitive to natural rubber latex Eliminates glove powder aerosolization Safe for food contact applications Glove Material- Vinyl Packaged Sterile- No Size Type- Medium Length- 12
Catalog No. 50-000-23021