Learn More
American Radiolabeled Chemicals Inc GLP-1 125I 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARI 015250UC
Encompass
GLP-1 (7-36), Tyr-125I 10uCi in buffered solution, 0.08 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~3298
Catalog No. 50-901-042
Only null left
Add to Cart
Request a Quote