Learn More
Medchemexpress LLC HY-P0055 1mg Medchemexpress, GLP-1(7-37) CAS:106612-94-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP00551MG
Encompass_Preferred
Medchemexpress, HY-P0055 1mg GLP-1(7-37) CAS:106612-94-6 GLP-1(7-37) is an intestinal insulinotropic hormone that augments glucose induced insulin secretion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4003