Learn More
Alpco Ltd GLP-1 (7-36) Specific ELISA
SDP
Supplier:  Alpco Ltd 43GP1HUE01
Encompass_Preferred
The highly sensitive GLP-1 ELISA (Active 7-36) is for the quantification of bioactive GLP-1 (7-36) levels in human EDTA-plasma.
Superior analytical sensitivity
0.05 pmol/L
Broad dynamic range
0.64–48 pmol/L
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-297