Learn More
IBL America Glucagon-like Peptide-1 (GLP-1), Active Form (h/m/r) ELISA
Supplier:  IBL America 27784
Encompass_Preferred
Solid phase sandwich ELISA for the determination of active form of GLP-1 (GLP-1 (7-36) amide and GLP-1 (7-37)) in EDTA plasma. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0372