Learn More
IBL America Glucagon-like Peptide-1 (GLP-1), Active Form (High Sensitivity)(h/m/r) ELISA
Supplier:  IBL America 27700
Encompass_Preferred
Solid phase sandwich enzyme-linked immunosorbent assay (ELISA) for the determination of GLP-1 (Active Form) in human, mouse and rat EDTA-plasma, and cell culture supernatant. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0337