Learn More
Cayman Chemical GLP-11-32bulfrogtrIf 1mg
SDP
Supplier:  Cayman Chemical 407181 MG
Encompass_Preferred
An endogenous incretin hormone; increases intracellular cAMP levels using HEK293 cells expressing human GLP-1R (EC50 = 0.051 nM); reduces blood glucose levels in an OGTT in fasted mice at 30 mg/kg
Catalog No. 50-294-2517