Learn More
Cayman Chemical GLP-128-36 amIdetrIfluor 10mg
SDP
Supplier:  Cayman Chemical 4124910 MG
Encompass_Preferred
A peptide and an active metabolite of GLP-1 (7-36) amide; inhibits hydrogen peroxide-induced decreases in ATP levels and t-BHP-induced increases in ROS levels in primary mouse hepatocytes and H-4-II-E cells at 100 nM; reduces body weight gain, the ratio of weight gain to food intake, and hepatic gluconeogenesis in a mouse model of HFD-induced obesity at 18.5 nmol/kg per day; decreases plasma levels of glucose and insulin and hepatic triglyceride levels in a mouse model of VHFD-induced obesity when infused at 18.5 nmol/kg per day
Catalog No. 50-294-4846