Learn More
Cayman Chemical GLP-17-36 amIdetrIfluoro 1mg
SDP
Supplier:  Cayman Chemical 150691 MG
Encompass_Preferred
A peptide hormone released from intestinal L-cells upon nutrient consumption that binds the GLP-1 receptor (pIC50 = 8.5) in the pancreas and displays various antidiabetic effects
Catalog No. 50-297-0649