Learn More
Medchemexpress LLC GLP-1R agonist 4 | 2401894-26-4 | 98.26% | C32H30ClF2N3O5 | 50 MG

Supplier: Medchemexpress LLC HY14403550MG
GLP-1R agonist 4 is a potent agonist of GLP-1R, an intestinal hypoglycemic hormone secreted by L-cells in the lower gastrointestinal tract. This compound has potential for research related to diabetes, as extracted from patent WO2019239319A1, compound 96. It is intended for research use only.
- Potent agonist of GLP-1R
- Potential for diabetes research
- Available in various quantities
- Appearance: Solid, white to off-white
- Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO (100 mg/mL)
- Protocols available for in vivo dissolution using various co-solvents
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.