Learn More
Cayman Chemical GLP-1RhumnrecombInta 100ug
SDP
Supplier:  Cayman Chemical 40466100 UG
Encompass_Preferred
Source: Recombinant C-terminal human IgG1 Fc-tagged GLP-1R expressed in HEK293 cells • Amino acids: 24-145 • MW: 41 kDa
Catalog No. 50-294-3715