Learn More
Medchemexpress LLC Glp-26 | 2133017-36-2 | 98.4% | 373.35 | C19H17F2N3O3 | 10MG

Supplier: Medchemexpress LLC HY12461410MG
GLP-26 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator for research use that inhibits viral DNA replication, reduces covalently closed circular DNA (cccDNA), and shows activity in multiple in vitro HBV models. The compound contains an alkyne group enabling click chemistry and is provided with recommended solubility and storage conditions for laboratory use.
- Potent HBV capsid assembly modulator with low-nanomolar activity.
- Reduces cccDNA and inhibits HBV replication in cellular assays.
- Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition.
- High purity (98.4%) suitable for research applications.
- Soluble in DMSO and formulatable for in vitro studies.
- Stable as powder under recommended frozen storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.