Learn More
Medchemexpress LLC Glp-26 | 2133017-36-2 | 98.4% | 373.35 | C19H17F2N3O3 | 10MM 1ML

Supplier: Medchemexpress LLC HY12461410MM 1ML
GLP-26 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator that inhibits HBV DNA replication (IC50 = 3 nM) and reduces covalently closed circular DNA (cccDNA) by >90% at 1 μM. The compound contains an alkyne functional group enabling copper-catalyzed azide-alkyne cycloaddition and is provided as a 10 mM solution in DMSO.
- Inhibits HBV DNA replication with IC50 of 3 nM.
- Reduces cccDNA levels by more than 90% at 1 μM.
- Contains an alkyne group suitable for click chemistry.
- Supplied as a 10 mM solution in DMSO (1 mL).
- Purity 98.4% as reported by the manufacturer.
- Molecular formula C19H17F2N3O3 and molecular weight 373.35 g·mol⁻1.
- Store powder at -20°C; store in solvent at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.