Learn More
Medchemexpress LLC Glp-26 | 2133017-36-2 | 98.4% | 373.35 g/mol | C19H17F2N3O3 | 100MG

Supplier: Medchemexpress LLC HY124614100MG
GLP-26 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator used in antiviral research. It inhibits HBV DNA replication (IC50 = 3 nM in Hep AD38 cells), reduces covalently closed circular DNA (cccDNA) by >90% at 1 μM, and contains an alkyne functionality enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC). The compound is supplied as a solid with high reported purity and documented solubility and storage recommendations for in vitro and in vivo use.
- Potent HBV inhibition (IC50 = 3 nM).
- Reduces cccDNA by >90% at 1 μM.
- Contains an alkyne group for click chemistry (CuAAC).
- High reported purity for reproducible results.
- Soluble in DMSO (125 mg/mL) with documented in vivo formulation protocols.
- Documented storage recommendations for powder and solutions to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.