Learn More
Medchemexpress LLC HY-100506 1mg Medchemexpress, GLPG0187 CAS:1320346-97-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1005061MG
Encompass_Preferred
Medchemexpress, HY-100506 1mg GLPG0187 CAS:1320346-97-1 GLPG0187 is a broad spectrum integrin receptor antagonist with antitumor activity; inhibits αvβ1-integrin with an IC50 of 1.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8625