Learn More
Medchemexpress LLC HY-18300 100mg , GLPG0634 Filgotinib CAS:1206161-97-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY18300100MG
Encompass_Preferred
Medchemexpress, HY-18300 100mg GLPG0634 Filgotinib CAS:1206161-97-8 Formula:C21H23N5O3S IC50: 10 nM (JAK1), 28 nM (JAK2), 810 nM (JAK3), 116 nM (Tyk2) Purity:98% GLPG0634 is a selective JAK1 inhibitor with IC 50 of 10 nM, 28 nM, 810 nM, and 116 nM for JAK1, JAK2, JAK3, and TYK2, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2883