Learn More
Medchemexpress LLC Glpg1837 (abbv-974) | 1654725-02-6 | 99.9% | 348.42 | 10 UG
SDP
Supplier:  Medchemexpress LLC HY11109910MG
Encompass_Preferred
GLPG1837 is a potent and reversible CFTR potentiator, with EC50s of 3 nM and 339 nM for F508del and G551D CFTR, respectively. It increases the conductivity of the G551D CFTR channel with an obtained potency of 181 nM.
  • Potent and reversible CFTR potentiator.
  • EC50s of 3 nM and 339 nM for F508del and G551D CFTR, respectively.
  • Increases the conductivity of the G551D CFTR channel with an obtained potency of 181 nM.
Catalog No. 50-003-43912