Learn More
Medchemexpress LLC Gls-1-in-1 | 00-00-0 | 99.6% | 460.57 g·mol⁻¹ | C26H25FN4OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1446665MG
Encompass_Preferred
GLS-1-IN-1 (compound 1d) is a small-molecule glutaminase 1 (GLS-1) inhibitor provided for research use. It demonstrates inhibitory activity against Hep G2, MCF-7, and MCF-10A cell lines and is supplied as a high-purity compound for in vitro studies.
  • Demonstrates inhibitory activity against Hep G2, MCF-7, and MCF-10A cells.
  • High purity suitable for research applications.
  • Chemical formula C26H25FN4OS and molecular weight 460.57 g·mol⁻¹.
  • Available in small milligram quantities for laboratory testing.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-21290