Learn More
Enzo Life Sciences Glu-Glu monoclonal antibody (Glu-Glu) (0.5ml)
SDP
Supplier:  Enzo Life Sciences ENZABS1610500
Encompass_Preferred
Clone: Glu-Glu. Isotype: IgG1. Immunogen: Peptide sequence CEEEEYMPME derived from the polyoma virus medium T antigen. Applications: IF, IP, WB. Purity: Purified by affinity chromatography. Formulation: Liquid. Phosphate-buffered solution + 0.03% Thimerosal.
Catalog No. 50-201-2719