Learn More
CPC Scientific H-Glu-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific FIBN004B
Encompass
ONE-LETTER SEQUENCE: EGVNDNEEGFFSARMOLECULAR FORMULA: C66H95N19O26MOLECULAR WEIGHT:1570.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [103213-49-6], RESEARCH AREA: Inflammation
Catalog No. 50-210-9278