Learn More
Medchemexpress LLC (Glu17,21,24)-Osteocalcin (1-49) (human) | 1927927-11-4 | 98.3% | 5797.40 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP45881MG
Encompass_Preferred
(Glu17,21,24)-Osteocalcin (1-49) (human) is a bone-specific protein involved in bone formation and glucose metabolism. It is intended for research use only and is not validated for medical applications.
  • Bone-specific protein
  • Involved in bone formation
  • Involved in glucose metabolism
  • Targets GLUT pathway
  • Acts on membrane transporter/ion channels
Catalog No. 50-003-98827