Learn More
Sigma Aldrich Fine Chemicals Biosciences Glucagon 95 HPLC powder sy1MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G1774
Encompass
Glucagon is synthesized from proglucagon and is 29-amino acid peptide hormone. Proglucagon gene is mapped to human chromosome 2q24.2.
Catalog No. 50-290-7292