Learn More
Mercodia Inc GLUCAGON ELISA
SDP
Supplier:  Mercodia Inc 10127101
Encompass_Preferred
The Mercodia Glucagon ELISA is a highly specific and sensitive assay for measuring glucagon in plasma, serum, and cell culture media. The highly specific monoclonal antibodies used in this assay enables accurate determination of glucagon without significant cross-reactivity to other circulating pro-glucagon derived peptides, like for instance glicentin.
Catalog No. NC0524433