Learn More
Selleck Chemical LLC Glucagon hydrochloride P1056-1G
SDP
Supplier:  Selleck Chemical LLC P10561G
Encompass_Preferred
Glucagon hydrochloride stimulates glycogenolysis in the liver used as an antihypoglycemic and as an adjunct in gastrointestinal radiography
Catalog No. 50-000-42666