Learn More
Medchemexpress LLC Glucagon-like peptide 1 (1-37), human | 87805-34-3 | 99.7% | 4169.48 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP114510MG
Encompass_Preferred
Glucagon-like peptide 1 (1-37), human is a highly potent agonist of the GLP-1 receptor. It is produced by the posttranslational processing of proglucagon and acts as a regulator of various homeostatic events.
  • Highly potent agonist of the GLP-1 receptor.
  • More stable than GLP-1(7-37), with 94.7% remaining after 4-hour mouse serum exposure.
  • Decreases glycemic excursion in a dose-dependent manner.
  • Significantly reduces blood glucose levels at 15 and 30 minutes compared to the control group.
Catalog No. 50-004-24747