Learn More
Medchemexpress LLC Glucoerucin potassium | 15592-37-7 | 1 MG

Supplier: Medchemexpress LLC HYN84451MG
Glucoerucin potassium is an aryl hydrocarbon receptor antagonist and is the 4-methylthio-butyl glucosinolate found in the seeds of Eruca sativa Mill. This compound is intended for research use only and not for patient administration.
- Aryl hydrocarbon receptor antagonist
- Derived from Eruca sativa Mill seeds
- Purity of 99.73%
- Molecular weight of 459.60
- Appears as a white to off-white solid
- Suitable for research applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.