Learn More
Medchemexpress LLC Gluconate-13C6 sodium | 2687960-00-3 | 100.0% | 224.09 | 13C6H11NaO7 | 100mg
SDP
Supplier:  Medchemexpress LLC HYW768336100MG
Encompass_Preferred
Gluconate sodium-13C6 (D-Gluconic acid sodium salt-13C6) is the 13C-labeled Gluconate sodium (HY-B1092A) Gluconate sodium (D-Gluconic acid sodium salt) is an orally active glucose derivative Gluconate sodium reduces nitric oxide and inflammatory cytokines (IL-1 and IL-6) Gluconate sodium inhibits ERK phosphorylation Gluconate sodium has antioxidant and antiplatelet activation activities Gluconate sodium has antitumor activity against colorectal cancer Gluconate sodium improves osteoarthritis intestinal damage and acute lung injury[1][2][3][4][5][6][7]
Catalog No. 50-002-69523