Learn More
TARGETMOL CHEMICALS INC Glucoraphanin 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ02125MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Glucoraphanin, the precursor of the widely extolled chemopreventive agent sulforaphane found in broccoli, has antioxidant activity.
Catalog No. 50-287-7315