Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucosamine | 3416-24-8 | ≥98.0% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYB112550MG

Encompass_Preferred

Glucosamine (D-Glucosamine; Chitosamine) is an amino sugar that serves as a precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is a natural component of glycosaminoglycans found in the cartilage matrix and synovial fluid. When administered externally, it has pharmacological effects on osteoarthritic cartilage and chondrocytes.

  • Appearance: Solid, white to off-white
  • Molecular weight: 179.17
  • Formula: C6H13NO5
  • Structure classification: Saccharides, monosaccharides
  • Initial source: Endogenous metabolite, human gut microbiota metabolites, microorganisms

Catalog No. 50-004-13073


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.