Learn More
Crescent Chemical Co Inc GLUCOSE ANHYDROUS
SDP
Supplier:  Crescent Chemical Co Inc SE22700.02
Encompass_Preferred
Glucose anhydrous analytical grade Assay (titr.)Water (loss on drying) min. 99.5 %max. 0.5 % Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 22700.02 CAS 50-99-7
Catalog No. 50-247-634