Learn More
Fisher Science Education™ Glucose Solution, 0.1%

$21.15
Chemical Identifiers
| CAS | 50-99-7 , 7732-18-5 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.156 |
| InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
| Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
| PubChem CID | 107526 |
| ChEBI | CHEBI:42758 |
| IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
| SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
Description
This product is for educational use - no C of A is available.
Chemical Identifiers
| 50-99-7 , 7732-18-5 | |
| 180.156 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| CHEBI:42758 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
| C6H12O6 | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| 107526 | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Specifications
| 50-99-7 , 7732-18-5 | |
| C6H12O6 | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
| 107526 | |
| Glucose Solution, 0.1% |
| 0.1% w/v | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| 180.156 | |
| CHEBI:42758 |
Safety and Handling
ShelfLife : 2 years
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.