Learn More
Medchemexpress LLC Glucosylceramide synthase-IN-3 | 3029827-91-3 | 99.1% | C21H20FN3O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14427010MG
Encompass_Preferred
Glucosylceramide synthase-IN-3 is a high-purity small-molecule inhibitor of glucosylceramide synthase (GCS) used in research on sphingolipid metabolism and neuronal signaling. Supplied as a stable solid with recommended storage information, it is appropriate for biochemical and cellular assays requiring selective GCS modulation.
  • High purity and enantiomeric excess suitable for research-grade experiments.
  • Targeted inhibition of glucosylceramide synthase (GCS) for mechanistic studies.
  • Available in multiple small-scale solid and solution formats for flexibility.
  • Recommended storage conditions provided for long-term stability.
  • Includes supporting datasheet and safety documentation.
Catalog No. 50-002-58396