Learn More
Medchemexpress LLC Glucosylsphingosine | 52050-17-6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN7745S1MG
Encompass_Preferred
Glucosylsphingosine (lyso-Gb1) is a deacylated form of glucosylceramide exhibiting neurotoxicity. It acts as a noncompetitive inhibitor of glucocerebrosidase with a Ki value of 30 μM. This compound is promising for research into Krabbe and Gaucher diseases.
  • Acts as a noncompetitive inhibitor of glucocerebrosidase.
  • Promising for research of Krabbe and Gaucher disease.
  • Induces LA-N-2 cell shriveling and suppresses neurite outgrowth in vitro.
  • Reduces activities of lysosomal enzymes including glucocerebrosidase, sphingomyelinase, and β-galactosidase.
  • Causes cell death in MCF7 cells independently of apoptosis, ferroptosis, and necroptosis.
  • Activates the cAMP signaling pathway.
  • Induces a visceral inflammatory response in mice.
  • Accumulates in peripheral tissues and causes hematological symptoms in mice.
Catalog No. 50-004-17682