Learn More
Medchemexpress LLC Glucosylsphingosine | 52050-17-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN77455MG
Encompass_Preferred
Glucosylsphingosine (lyso-Gb1) is a deacylated form of glucosylceramide that exhibits neurotoxicity. It also functions as a noncompetitive inhibitor of glucocerebrosidase with a Ki value of 30 μM. This compound shows promise for research related to Krabbe and Gaucher disease.
  • Exhibits neurotoxicity.
  • Functions as a noncompetitive inhibitor of glucocerebrosidase with a Ki value of 30 μM.
  • Shows promise for research of Krabbe and Gaucher disease.
Catalog No. 50-004-17222