Learn More
Medchemexpress LLC Glucosylsphingosine | 52050-17-6 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYN7745S1500UG
Encompass_Preferred
Glucosylsphingosine (lyso-Gb1) is a deacylated form of glucosylceramide that exhibits neurotoxicity. It acts as a noncompetitive inhibitor of glucocerebrosidase and is relevant for research in Krabbe and Gaucher diseases.
  • Deacylated form of glucosylceramide
  • Exhibits neurotoxicity
  • Noncompetitive inhibitor of glucocerebrosidase
  • Used in research for Krabbe and Gaucher diseases
  • Suppresses neurite outgrowth
  • Reduces activity of lysosomal enzymes
  • Induces cell death in MCF7 cells
  • Activates cAMP signaling pathway
  • Induces visceral inflammatory response in mice
  • Causes hematological symptoms and tissue inflammation in mice
Catalog No. 50-004-19636