Learn More
Medchemexpress LLC Glucosylsphingosine | 52050-17-6 | MFCD07374303 | 1 MG

Supplier: Medchemexpress LLC HYN77451MG
Glucosylsphingosine (lyso-Gb1) is a deacylated form of glucosylceramide with neurotoxicity and is also a noncompetitively inhibitor of glucocerebrosidase with a Ki value of 30 μM. It is promising for research of Krabbe and Gaucher disease. It is supplied as a white to off-white solid with a high purity of 99.68%.
- Deacylated form of glucosylceramide.
- Exhibits neurotoxicity.
- Noncompetitive inhibitor of glucocerebrosidase.
- Potential for research in Krabbe disease.
- Potential for research in Gaucher disease.
- High purity of 99.68%.
- Supplied as a white to off-white solid.
- Store at 4°C, protected from light.
- Stable in solvent at -80°C for 6 months, or -20°C for 1 month (protected from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.