Learn More
Medchemexpress LLC Glucosyringic acid | 33228-65-8 | 5 MG

Supplier: Medchemexpress LLC HYN39535MG
Glucosyringic acid is a compound isolated from *Saxifraga Montana H.*. It is intended for research use only and is not sold to patients.
- Purity: 99.62%
- Molecular weight: 360.31
- Formula: C15H20O10
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants (Saxifragaceae, *Saxifraga sinomontana J. T. Pan & Gornall*)
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.