Learn More
Medchemexpress LLC Glucotropaeolin potassium | 5115-71-9 | 1 MG

Supplier: Medchemexpress LLC HYN43211MG
Glucotropaeolin potassium (Benzylglucosinolate potassium) is a glucosinolate found in cruciferous vegetables that causes a moderate decrease in spontaneous DNA damage in animals.
- Purity: 99.40%
- For research use only
- Molecular weight: 447.52
- Formula: C14H18KNO9S2
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants other families
- Shipping at room temperature in continental US; may vary elsewhere.
- Storage at -20°C, protected from light. In solvent: -80°C for 6 months; -20°C for 1 month (protected from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.