Learn More
Medchemexpress LLC Glucotropaeolin potassium | 5115-71-9 | MFCD00153006 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN43215MG
Encompass_Preferred
Glucotropaeolin potassium is a glucosinolate naturally found in cruciferous vegetables. This compound has been observed to cause a moderate decrease in spontaneous DNA damage in animals and is intended for various research applications, particularly in studies involving DNA synthesis and damage.
  • Naturally found in cruciferous vegetables
  • Causes a moderate decrease in spontaneous DNA damage in animals
  • Intended for research use only
  • Classified as an alkaloid and glucosinolate
Catalog No. 50-004-20787