Learn More
Medchemexpress LLC Glucovanillin | 494-08-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN66675MG
Encompass_Preferred
Glucovanillin, found in vanilla, is a potential lipase inhibitor. It can be converted into vanillin through an enzyme-coupled process involving cell wall degradation and glucovanillin hydrolysis. It has the potential to inhibit the lipase activity of *Acinetobacter radioresistens*.
  • Potential lipase inhibitor
  • Purity 99.87%
  • Available in solid and solution forms
  • For research use only
  • Targets lipase
  • Functions via metabolic enzyme/protease pathway
Catalog No. 50-004-19315