Learn More
Cayman Chemical GluCys ammonium salt 50mg
SDP
Supplier:  Cayman Chemical 3056950 MG
Encompass_Preferred
An intermediate in GSH synthesis; formed by ATP-dependent condensation of cysteine and glutamate by γ-glutamylcysteine synthetase; an essential component of phytochelatins
Catalog No. 50-299-2184