Learn More
ABclonal Technology GLUD1/2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A21792100UL
Encompass_Preferred
The protein encoded by this gene is localized to the mitochondrion and acts as a homohexamer to recycle glutamate during neurotransmission The encoded enzyme catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate This gene is intronless
Catalog No. 50-000-06611