Learn More
World Precision Instrument Mini Glue Gun with Glue Sticks
SDP
Supplier:  World Precision Instrument 13316
Encompass
  • Bonds wood, glass, metals, and many plastics
  • Easy to use for bonding and large gap filling
  • UL approved
  • 110 V 60 Hz only
Catalog No. 50-821-802